4-Bromo-2,6-dimethylbenzoic acid 98%

Supplier: Ambeed
Warning

A141064-10G A141064-250MG A141064-1G A141064-25G A141064-5G
103802-942EA 31 USD
103802-942 77027-522 77027-524 77299-188 77316-070 77316-072
4-Bromo-2,6-dimethylbenzoic acid 98%
4-Bromo-2,6-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₉BrO₂
MW: 229.0706 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09965997
CAS Number: 74346-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR