4-Bromo-2,6-dimethylbenzaldehyde 97%

Supplier: Ambeed
Warning

A251122-1G A251122-25G A251122-5G A251122-100G A251122-100MG A251122-250MG
103811-964EA 36 USD
103811-964 103851-806 77091-174 77091-176 77091-178 77883-679 77883-680 77883-681
4-Bromo-2,6-dimethylbenzaldehyde 97%
4-Bromo-2,6-dimethylbenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO
Storage Temperature: Refrigerator
MDL Number: MFCD11227151
CAS Number: 5769-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR