4-Bromo-2,6-dimethoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A173613-10G A173613-250MG A173613-1G A173613-25G A173613-5G A173613-100G
103806-842EA 10 USD
103806-842 103845-142 76985-550 76985-552 76985-554 77082-394 77082-396 77082-398 77890-119
4-Bromo-2,6-dimethoxybenzaldehyde 97%
4-Bromo-2,6-dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₉BrO₃
Storage Temperature: Refrigerator
MDL Number: MFCD22192498
CAS Number: 1354050-38-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR