4-Bromo-2,6-diisopropylaniline 98%

Supplier: Ambeed
Warning

A522114-1G A522114-25G A522114-5G A522114-10G A522114-100G
103828-514EA 10 USD
103828-514 103839-624 76843-622 76843-624 76843-626 76878-236 77551-576
4-Bromo-2,6-diisopropylaniline 98%
4-Bromo-2,6-diisopropylaniline

Precaución: for Research Use Only

Formula: C₁₂H₁₈BrN
Flash Pt: 135.546 °C
Storage Temperature: Ambient
MDL Number: MFCD09743895
CAS Number: 80058-84-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR