4-Bromo-2,6-difluorophenol 97%

Supplier: Ambeed
Warning

A102127-100G A102127-10G A102127-25G A102127-500G A102127-1G A102127-5G
103796-308EA 6 USD
103796-308 103822-000 77148-254 77148-256 77148-258 77148-260 77282-908 77282-910
4-Bromo-2,6-difluorophenol 97%
4-Bromo-2,6-difluorophenol

Precaución: for Research Use Only

Formula: C₆H₃BrF₂O
Boiling Pt: 180.2 °C (760 mm Hg)
Melting Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00236213
CAS Number: 104197-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR