4-Bromo-2,6-difluorobenzoic acid 98%
Supplier: Ambeed
|
Warning
|
A148222-100G
A148222-10G
A148222-500G
A148222-25G
A148222-5G
103842-008EA
10
USD
103842-008
76768-062
76768-064
76768-066
77317-442
77317-444
4-Bromo-2,6-difluorobenzoic acid 98%
4-Bromo-2,6-difluorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃BrF₂O₂ Boiling Pt: 271.7 °C (760 mm Hg) Melting Pt: 201…203 °C Flash Pt: 118.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD03094085 CAS Number: 183065-68-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...