4-Bromo-2,6-difluorobenzaldehyde 97%

Supplier: Ambeed
Warning

A248582-25G A248582-5G A248582-100G A248582-10G A248582-500G A248582-1G
103811-826EA 8 USD
103811-826 103851-660 77091-006 77091-008 77288-946 77288-948 77288-950 77304-448
4-Bromo-2,6-difluorobenzaldehyde 97%
4-Bromo-2,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
Melting Pt: 76…81 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD03094459
CAS Number: 537013-51-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR