4-Bromo-2,6-difluoroaniline 98%
Supplier: Ambeed
|
Warning
|
A300171-1000G
A300171-25G
A300171-100G
A300171-10G
A300171-1G
A300171-500G
A300171-5G
103810-012EA
337
USD
103810-012
76838-102
76838-104
77289-958
77289-960
77289-962
77289-964
77289-966
4-Bromo-2,6-difluoroaniline 98%
4-Bromo-2,6-difluoroaniline
Precaución: for Research Use Only
Formula:
C₆H₄BrF₂N Boiling Pt: 188.2 °C (760 mm Hg) Melting Pt: 63…65 °C Flash Pt: 67.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013389 CAS Number: 67567-26-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...