4-Bromo-2,6-dichlorobenzonitrile 98%

Supplier: Ambeed
Warning

A214178-100MG A214178-250MG A214178-1G A214178-5G
103818-374EA 27 USD
103818-374 103849-082 76785-758 76785-760 77087-588 77087-590
4-Bromo-2,6-dichlorobenzonitrile 98%
4-Bromo-2,6-dichlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrCl₂N
Storage Temperature: Ambient
MDL Number: MFCD14702850
CAS Number: 99835-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR