4-Bromo-2,6-dichlorobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A344289-10G
A344289-1G
A344289-100G
103825-900EA
10
USD
103825-900
103836-880
76817-266
76839-928
77672-750
4-Bromo-2,6-dichlorobenzoic acid 97%
4-Bromo-2,6-dichlorobenzoic acid
Precaución: for Research Use Only
Formula:
C₇H₃BrCl₂O₂ MW: 269.90 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11227148 CAS Number: 232275-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...