4-Bromo-2,6-dichlorobenzaldehyde 98%

Supplier: Ambeed
Warning

A139805-100MG A139805-250MG A139805-1G
103802-778EA 27 USD
103802-778 76823-376 76823-378 77315-868
4-Bromo-2,6-dichlorobenzaldehyde 98%
4-Bromo-2,6-dichlorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O
MW: 253.9 g/mol
Boiling Pt: 296.0±40.0 °C (760 mm Hg)
Flash Pt: 132.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11227147
CAS Number: 111829-72-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR