4-Bromo-2,5-dimethylpyridine 95%

Supplier: Ambeed
Warning

A210669-100MG A210669-250MG A210669-1G
103818-032EA 318 USD
103818-032 103848-726 76814-540 76814-542 77303-140
4-Bromo-2,5-dimethylpyridine 95%
4-Bromo-2,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00234307
CAS Number: 17117-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR