4-Bromo-2,5-dimethoxybenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
A167297-1G
A167297-25G
A167297-5G
A167297-100G
A167297-10G
A167297-250MG
103806-160EA
6
USD
103806-160
103844-420
77081-428
77081-430
77081-432
77286-652
77286-654
77286-656
4-Bromo-2,5-dimethoxybenzaldehyde 95%
4-Bromo-2,5-dimethoxybenzaldehyde
Precaución: for Research Use Only
Formula:
C₉H₉BrO₃ MW: 245.08 g/mol Boiling Pt: 337.1 °C (760 mm Hg) Flash Pt: 157.7 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00156964 CAS Number: 31558-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...