4-Bromo-2,5-difluoropyridine 95%

Supplier: Ambeed
Warning

A140028-100MG A140028-5G
103802-816EA 162 USD
103802-816 76984-140 77315-924
4-Bromo-2,5-difluoropyridine 95%
4-Bromo-2,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrF₂N
MW: 193.98 g/mol
Boiling Pt: 178.6±35.0 °C
Flash Pt: 61.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19686165
CAS Number: 1349718-16-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR