4-Bromo-2,5-difluorobenzonitrile 97% (by GC)

Supplier: Ambeed
Warning

A190510-10G A190510-250MG A190510-1G A190510-25G A190510-5G A190510-100G
103816-300EA 17 USD
103816-300 103846-902 76974-840 76974-842 77084-704 77084-706 77084-708 77671-696
4-Bromo-2,5-difluorobenzonitrile 97% (by GC)
4-Bromo-2,5-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₂BrF₂N
Storage Temperature: Ambient
MDL Number: MFCD11847049
CAS Number: 133541-45-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR