4-Bromo-2,5-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A148232-10G A148232-250MG A148232-1G A148232-5G
103803-894EA 12 USD
103803-894 103842-010 76871-514 76871-516 77317-446 77317-448
4-Bromo-2,5-difluorobenzaldehyde 98%
4-Bromo-2,5-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 220.999 g/mol
Boiling Pt: 237 °C (760 mm Hg)
Flash Pt: 97 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847047
CAS Number: 357405-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR