4-Bromo-2,5-dichloropyridine 95%

Supplier: Ambeed
Warning

A381558-250MG A381558-5G A381558-1G A381558-10G
103826-702EA 35 USD
103826-702 103837-736 76788-774 76841-052 77306-924 77668-762
4-Bromo-2,5-dichloropyridine 95%
4-Bromo-2,5-dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrCl₂N
MW: 226.89 g/mol
Boiling Pt: 254 °C (760 mm Hg)
Melting Pt: 60…61 °C
Flash Pt: 108 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12911586
CAS Number: 1184917-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR