4-Bromo-2,3-dimethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,3-lutidine

A286447-100MG A286447-10G A286447-25G A286447-1G A286447-5G A286447-250MG
103813-712EA 18 USD
103813-712 103834-848 76816-462 76816-464 76816-466 76837-382 76837-384 77672-480
4-Bromo-2,3-dimethylpyridine 98%
4-Bromo-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈BrN
MW: 186.049 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18253389
CAS Number: 259807-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR