4-Bromo-2,3-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A216934-100MG A216934-10G A216934-250MG A216934-1G A216934-25G A216934-5G
103818-588EA 13 USD
103818-588 103849-314 76975-720 76975-722 76975-724 77087-922 77087-924 77087-926
4-Bromo-2,3-difluorobenzaldehyde 98%
4-Bromo-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MW: 221.00 g/mol
Boiling Pt: 239.8 °C (760 mm Hg)
Melting Pt: 63…67 °C
Flash Pt: 98.864 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09258953
CAS Number: 644985-24-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR