4-Bromo-2,2'-bipyridyl 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,2'-bipyridine

A100059-5G A100059-1G A100059-250MG A100059-100MG
103795-224EA 95 USD
103795-224 103820-946 77296-556 77282-414 77282-416 77614-722
4-Bromo-2,2'-bipyridyl 97%
4-Bromo-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₇BrN₂
MW: 235.08 g/mol
Boiling Pt: 325.2 °C (760 mm Hg)
Flash Pt: 150.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09910216
CAS Number: 14162-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR