4-Bromo-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-α,α,α-trifluoro-2-picoline

A603554-1G A603554-5G A603554-10G A603554-25G
103828-990EA 10 USD
103828-990 103840-132 76844-290 76844-292 77292-806 77307-922
4-Bromo-2-(trifluoromethyl)pyridine 98%
4-Bromo-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 225.994 g/mol
Boiling Pt: 167.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD07774089
CAS Number: 887583-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR