4-Bromo-2-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

A189909-100G A189909-100MG A189909-10G A189909-250MG A189909-1G A189909-25G A189909-5G
103816-234EA 8 USD
103816-234 103846-830 76998-932 76998-934 76998-936 76998-938 77084-606 77084-608 77084-610
4-Bromo-2-(trifluoromethyl)benzoic acid 97%
4-Bromo-2-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄BrF₃O₂
Boiling Pt: 275.2 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07787468
CAS Number: 320-31-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR