4-Bromo-2-(hydroxymethyl)benzoic acid 95%

Supplier: Ambeed
Warning

A549506-250MG A549506-100MG
103828-684EA 107 USD
103828-684 103839-808 77017-652 77891-105
4-Bromo-2-(hydroxymethyl)benzoic acid 95%
4-Bromo-2-(hydroxymethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₇BrO₃
Boiling Pt: 383 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11977384
CAS Number: 670256-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR