3-(Difluoromethoxy)-4-fluorobromobenzene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Bromo-α,α,2-trifluoroanisole, 4-Bromo-2-(1,1-difluoromethoxy)-1-fluorobenzene, 4-Bromo-2-(difluoromethoxy)-1-fluorobenzene
A234747-10G
A234747-1G
A234747-5G
103810-972EA
32
USD
103810-972
103850-728
76786-392
77089-726
77303-992
3-(Difluoromethoxy)-4-fluorobromobenzene 97%
3-(Difluoromethoxy)-4-fluorobromobenzene
Precaución: for Research Use Only
Formula:
C₇H₄BrF₃O MW: 241.005 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11036068 CAS Number: 954235-78-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...