5-Bromo-2-nitrobenzyl bromide 97%
Supplier: Ambeed
|
Danger
|
A158980-250MG
A158980-1G
103805-206EA
139
USD
103805-206
103843-418
77011-130
77319-280
5-Bromo-2-nitrobenzyl bromide 97%
5-Bromo-2-nitrobenzyl bromide
Precaución: for Research Use Only
Formula:
C₇H₅Br₂NO₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD11848689 CAS Number: 35287-42-4 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...