4-Bromo-1-phenyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A257688-1G A257688-25G A257688-5G
103812-332EA 25 USD
103812-332 103852-200 77091-722 77091-724 77091-726
4-Bromo-1-phenyl-1H-pyrazole 97%
4-Bromo-1-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Boiling Pt: 296.3 °C (760 mm Hg)
Flash Pt: 133 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00234505
CAS Number: 15115-52-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR