4-Bromo-1-naphthoic acid 97%

Supplier: Ambeed
Warning

A244892-100G A244892-10G A244892-500G A244892-1G A244892-25G A244892-5G
103811-608EA 6 USD
103811-608 103851-442 77013-850 77013-852 77013-854 77090-660 77304-338 77304-340
4-Bromo-1-naphthoic acid 97%
4-Bromo-1-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇BrO₂
Boiling Pt: 408.8 °C (760 mm Hg)
Melting Pt: 198…200 °C
Flash Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD00094221
CAS Number: 16650-55-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR