4-Bromo-1-naphthaldehyde 97%
Supplier: Ambeed
|
Warning
|
A118671-100MG
A118671-10G
A118671-250MG
A118671-25G
A118671-1G
A118671-5G
103799-092EA
8
USD
103799-092
76983-098
76983-100
76983-102
76983-104
77311-762
77311-764
4-Bromo-1-naphthaldehyde 97%
4-Bromo-1-naphthaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₇BrO Melting Pt: 250 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11101026 CAS Number: 50672-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...