4-Bromo-1-methylpyridin-2(1H)-one 97%

Supplier: Ambeed
Warning

A223605-10G A223605-1G A223605-25G A223605-5G A223605-250MG
103819-046EA 9 USD
103819-046 103849-926 76786-068 77088-548 77088-550 77303-654 77593-456
4-Bromo-1-methylpyridin-2(1H)-one 97%
4-Bromo-1-methylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₆BrNO
MW: 188.0200 g/mol
Boiling Pt: 258 °C (760 mm Hg)
Melting Pt: 89…92 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11226870
CAS Number: 214342-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR