4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

A329799-1G A329799-10G A329799-250MG A329799-25G
103825-538EA 11 USD
103825-538 103836-496 76839-470 77290-404 77290-406 77290-408
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole 97%
4-Bromo-1-methyl-3-(trifluoromethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₅H₄BrF₃N₂
Boiling Pt: 200.4 °C (760 mm Hg)
Flash Pt: 75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115415
CAS Number: 497832-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR