4-Bromo-1-methyl-1H-pyrazole-5-carbonitrile 97%

Supplier: Ambeed
Warning

A223411-1G A223411-5G A223411-100MG A223411-250MG
103819-034EA 119 USD
103819-034 103849-914 77088-532 77088-534 77192-948 77192-950
4-Bromo-1-methyl-1H-pyrazole-5-carbonitrile 97%
4-Bromo-1-methyl-1H-pyrazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₄BrN₃
MW: 186.01 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00463999
CAS Number: 327099-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR