4-Bromo-1-methyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

A283956-5G A283956-100G A283956-10G A283956-1G
103834-760EA 7 USD
103834-760 76837-292 77114-110 77114-112 77114-114
4-Bromo-1-methyl-1H-pyrazole 97%
4-Bromo-1-methyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₄H₅BrN₂
MW: 161.00 g/mol
Boiling Pt: 185…188 °C (760 mm Hg)
Flash Pt: 93.3 °C
Storage Temperature: Ambient
MDL Number: MFCD02179565
CAS Number: 15803-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR