3-Amino-4-bromo-1-methyl-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-1-methyl-1H-pyrazol-3-amine, 3-Amino-4-bromo-1-methyl-1H-pyrazole, 3-Amino-4-bromo-1-methylpyrazole
A154562-100G
A154562-10G
A154562-250MG
A154562-1G
A154562-25G
A154562-5G
103804-654EA
23
USD
103804-654
103842-844
77273-096
77273-098
77273-100
77318-574
77318-576
77318-578
3-Amino-4-bromo-1-methyl-1H-pyrazole 97%
3-Amino-4-bromo-1-methyl-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₄H₆BrN₃ MW: 176.02 g/mol Boiling Pt: 274.6 °C (760 mm Hg) Melting Pt: 90 °C Flash Pt: 119.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00663065 CAS Number: 146941-72-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...