4-Bromo-2-nitroanisole 98%

Supplier: Ambeed
Warning

A106601-100G A106601-10G A106601-25G A106601-5G
103822-804EA 6 USD
103822-804 76765-504 76765-506 76765-508 77283-488
4-Bromo-2-nitroanisole 98%
4-Bromo-2-nitroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
Boiling Pt: 303.3 °C (760 mm Hg)
Melting Pt: 85…100 °C
Flash Pt: 137.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00055529
CAS Number: 33696-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR