4-Bromo-2-methylanisole 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-2-methoxytoluene, 1-Bromo-4-methoxy-3-methylbenzene, 4-Bromo-1-methoxy-2-methylbenzene

A136397-100G A136397-10G A136397-500G A136397-5G
77285-524EA 82.4 USD
77285-524 77285-526 77285-528 77315-188
4-Bromo-2-methylanisole 98%
4-Bromo-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.06 g/mol
Boiling Pt: 226.1 °C (760 mm Hg)
Melting Pt: 66…69 °C
Storage Temperature: Ambient
MDL Number: MFCD01321139
CAS Number: 14804-31-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR