4-Bromo-1-isopropyl-1H-imidazole 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1-(prop-2-yl)-1H-imidazole

A210498-250MG A210498-100MG
103818-006EA 125 USD
103818-006 103848-702 77612-070 77892-318
4-Bromo-1-isopropyl-1H-imidazole 97%
4-Bromo-1-isopropyl-1H-imidazole

Precaución: Sealed in dry, at room temperature.

Formula: C₆H₉BrN₂
MW: 189.053 g/mol
Boiling Pt: 268.7±13.0 °C (760 mm Hg)
Flash Pt: 116.3 °C
Storage Temperature: Ambient
MDL Number: MFCD22372008
CAS Number: 623577-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR