5-Bromo-2-iodoanisole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-1-iodo-2-methoxybenzene
A105773-100G
A105773-10G
A105773-250MG
A105773-1G
A105773-5G
A105773-25G
103796-938EA
12
USD
103796-938
103822-680
76970-422
76970-424
76970-426
77309-544
77309-546
77610-832
5-Bromo-2-iodoanisole 98%
5-Bromo-2-iodoanisole
Precaución: for Research Use Only
Formula:
C₇H₆BrIO MW: 312.9300 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD11847482 CAS Number: 791642-68-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...