4-Bromo-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A280797-1G A280797-250MG
103813-524EA 124 USD
103813-524 103853-472 76837-164 77672-448
4-Bromo-1H-pyrrolo[3,2-c]pyridine 95%
4-Bromo-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.032 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08690131
CAS Number: 1000342-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR