4-Bromo-1H-pyrrolo[2,3-c]pyridine 98%

Supplier: Ambeed
Warning

A364964-1G A364964-250MG
103826-374EA 23 USD
103826-374 103837-394 76840-628 77613-080
4-Bromo-1H-pyrrolo[2,3-c]pyridine 98%
4-Bromo-1H-pyrrolo[2,3-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrN₂
Boiling Pt: 337.6 °C (760 mm Hg)
Melting Pt: 188…189 °C
Flash Pt: 157.986 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11847768
CAS Number: 69872-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR