4-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95%
Supplier: Ambeed
|
Warning
|
A261598-100MG
A261598-1G
103812-530EA
425
USD
103812-530
76874-874
77092-028
4-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one 95%
4-Bromo-1H-pyrrolo[2,3-c]pyridin-2(3H)-one
Precaución: for Research Use Only
Formula:
C₇H₅BrN₂O MW: 213.03 g/mol Boiling Pt: 357.1 °C (760 mm Hg) Flash Pt: 169.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD12963348 CAS Number: 1190318-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...