4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 97%

Supplier: Ambeed
Warning

A119043-100MG A119043-250MG
103799-156EA 70 USD
103799-156 76983-110 76983-112
4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile 97%
4-Bromo-1H-pyrrolo[2,3-b]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.04 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11046238
CAS Number: 1159982-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR