4-Bromo-1H-pyrrole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A142133-100G A142133-10G A142133-250MG A142133-1G A142133-25G A142133-5G
103803-104EA 7 USD
103803-104 77285-790 77285-792 77285-794 77316-318 77316-320 77316-322
4-Bromo-1H-pyrrole-2-carboxylic acid 95%
4-Bromo-1H-pyrrole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂
MW: 189.9948 g/mol
Boiling Pt: 393 °C (760 mm Hg)
Melting Pt: 168…170 °C
Flash Pt: 191.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00832862
CAS Number: 27746-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR