4-Bromo-1H-pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

A137093-250MG A137093-1G A137093-25G A137093-10G
103802-402EA 36 USD
103802-402 76782-708 77298-884 77315-354 77871-654
4-Bromo-1H-pyrazolo[3,4-b]pyridine 97%
4-Bromo-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
Boiling Pt: 209 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10699404
CAS Number: 856859-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR