4-Bromo-1H-pyrazole-5-carbonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1H-pyrazole-3-carbonitrile

A178554-5G A178554-250MG A178554-10G
103807-352EA 8 USD
103807-352 103845-688 77083-120 77287-082 77748-166
4-Bromo-1H-pyrazole-5-carbonitrile 97%
4-Bromo-1H-pyrazole-5-carbonitrile

Precaución: for Research Use Only

Formula: C₄H₂BrN₃
MW: 171.98 g/mol
Boiling Pt: 412.4 °C (760 mm Hg)
Melting Pt: 154…158 °C
Flash Pt: 203.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00105683
CAS Number: 288246-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR