4-Bromo-1H-pyrazole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

A177229-1G A177229-5G A177229-10G A177229-250MG
103807-218EA 6 USD
103807-218 103845-546 77082-934 77082-936 77287-050 77287-052
4-Bromo-1H-pyrazole-3-carboxylic acid 98%
4-Bromo-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O₂
MW: 190.9828 g/mol
Boiling Pt: 440.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD01459882
CAS Number: 13745-17-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR