3-Amino-4-bromo-1H-pyrazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole
A228868-100G
A228868-10G
A228868-250MG
A228868-1G
A228868-25G
A228868-5G
A228868-500G
103819-424EA
9
USD
103819-424
103850-330
76976-066
76976-068
76976-070
77089-136
77089-138
77089-140
77881-590
3-Amino-4-bromo-1H-pyrazole 98%
3-Amino-4-bromo-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₃H₄BrN₃ MW: 161.99 Boiling Pt: 346.6 °C (760 mm Hg) Melting Pt: 133…135 °C Flash Pt: 163.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00082728 CAS Number: 16461-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...