4-Bromo-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A226005-10G A226005-250MG A226005-1G A226005-25G
103819-212EA 43 USD
103819-212 103850-114 77000-068 77000-070 77088-840 77303-756
4-Bromo-1H-indole-2-carboxylic acid 95%
4-Bromo-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
Boiling Pt: 470.9 °C (760 mm Hg)
Flash Pt: 238.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02664468
CAS Number: 16732-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR