4-Bromo-1H-indole-2-carbaldehyde 96%

Supplier: Ambeed
Warning

A118904-250MG A118904-1G A118904-5G
103799-128EA 296 USD
103799-128 76822-294 77297-636 77297-638
4-Bromo-1H-indole-2-carbaldehyde 96%
4-Bromo-1H-indole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.05 g/mol
Boiling Pt: 395.6±22.0 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD22035920
CAS Number: 1368231-70-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR