4-Bromoindole 97% (by HPLC)

Supplier: Ambeed
Warning

A119088-100G A119088-10G A119088-25G A119088-1G A119088-5G A119088-500G
103799-160EA 5 USD
103799-160 77233-988 77233-990 77233-992 77311-828 77311-830 77745-872
4-Bromoindole 97% (by HPLC)
4-Bromoindole

Precaución: for Research Use Only

Formula: C₈H₆BrN
Boiling Pt: 316.9 °C (760 mm Hg)
Melting Pt: 17 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00671502
CAS Number: 52488-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR