4-Bromo-1H-indazole-6-carbonitrile 95%

Supplier: Ambeed
Warning

A673397-1G A673397-100MG A673397-250MG A673397-5G
103840-508EA 194 USD
103840-508 76844-704 76992-536 76992-538 77308-098
4-Bromo-1H-indazole-6-carbonitrile 95%
4-Bromo-1H-indazole-6-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄BrN₃
MW: 222.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458975
CAS Number: 898746-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR